Assay Details
| Gene Symbol: Frs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144939.2 | NP_659188.1 | 4 - 5 | 489 | 54 | |
| GenBank mRNA | BC014819.1 | - | 4 - 5 | 489 | 4506982 | 54 |
More Information
Updated Mapping Information
Set Membership: |
|

Frs3
Genomic Map