Assay Details
| Gene Symbol: Eif2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145371.3 | NP_663346.1 | 6 - 7 | 593 | 70 | |
| GenBank mRNA | AK088066.1 | - | 6 - 7 | 592 | 70 | |
| AK133695.1 | - | 6 - 7 | 594 | 70 | ||
| BC003426.1 | - | 6 - 7 | 593 | 2615801 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif2b1
Genomic Map