Assay Details
| Gene Symbol: Lpcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145376.5 | NP_663351.3 | 4 - 5 | 603 | 74 | |
| GenBank mRNA | AB244717.2 | - | 4 - 5 | 603 | 74 | |
| AK046079.1 | - | 2 - 3 | 398 | 74 | ||
| AK046507.1 | - | 2 - 3 | 398 | 74 | ||
| AK079946.1 | - | 761 | 74 | |||
| AK081865.1 | - | 4 - 5 | 761 | 74 | ||
| AK132379.1 | - | 4 - 5 | 737 | 74 | ||
| AK155286.1 | - | 4 - 5 | 767 | 74 | ||
| AK169190.1 | - | 4 - 5 | 736 | 74 | ||
| AK170723.1 | - | 4 - 5 | 756 | 74 | ||
| AK171582.1 | - | 4 - 5 | 756 | 74 | ||
| BC005662.1 | - | 2 - 3 | 133 | 74 | ||
| BC066809.1 | - | 4 - 5 | 552 | 30475544 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046079.1, AK046507.1, AK079946.1, BC005662.1 transcripts.
Set Membership: |
|

Lpcat1
Genomic Map