Assay Details
Gene Symbol: Deptor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037937.3 | NP_001033026.1 | 6 - 7 | 929 | 68 | |
NM_145470.3 | NP_663445.2 | 6 - 7 | 1107 | 68 | ||
GenBank mRNA | AK145208.1 | - | 6 - 7 | 1001 | 68 | |
AK220300.1 | - | 6 - 7 | 941 | 68 | ||
BC004774.1 | - | 6 - 7 | 1112 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
