Assay Details
| Gene Symbol: Alg12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142357.1 | NP_001135829.1 | 3 - 4 | 388 | 87 | |
| NM_145477.2 | NP_663452.1 | 3 - 4 | 388 | 87 | ||
| GenBank mRNA | AJ429133.1 | - | 3 - 4 | 348 | 87 | |
| AK142322.1 | - | 3 - 4 | 389 | 87 | ||
| BC021379.1 | - | 3 - 4 | 361 | 5044319 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alg12
Genomic Map