Assay Details
| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145828.3 | NP_665827.2 | 2 - 3 | 714 | 84 | |
| GenBank mRNA | AJ291751.2 | - | 2 - 3 | 631 | 84 | |
| AK083482.1 | - | 2 - 3 | 683 | 84 | ||
| AK150521.1 | - | 2 - 3 | 667 | 84 | ||
| AK153332.1 | - | 2 - 3 | 667 | 84 | ||
| AK158056.1 | - | 2 - 3 | 714 | 84 | ||
| BC034082.1 | - | 1 - 2 | 233 | 4239646 | 84 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC034082.1 transcripts.
Set Membership: |
|

Xylt2
Genomic Map