Assay Details
| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145828.3 | NP_665827.2 | 6 - 7 | 1392 | 77 | |
| GenBank mRNA | AJ291751.2 | - | 6 - 7 | 1309 | 77 | |
| AK083482.1 | - | 6 - 7 | 1361 | 77 | ||
| AK086804.1 | - | 6 - 7 | 1324 | 77 | ||
| AK150521.1 | - | 6 - 7 | 1345 | 77 | ||
| AK153332.1 | - | 6 - 7 | 1345 | 77 | ||
| AK158056.1 | - | 6 - 7 | 1392 | 77 | ||
| BC034082.1 | - | 5 - 6 | 911 | 4239646 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xylt2
Genomic Map