Assay Details
Gene Symbol: Rnf144b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170643.1 | NP_001164114.1 | 1 - 2 | 119 | 80 | |
GenBank mRNA | AK040939.1 | - | 1 - 2 | 111 | 80 | |
AK052846.1 | - | 1 - 2 | 120 | 80 | ||
BB661852.1 | - | 1 - 2 | 120 | 80 | ||
BC025007.1 | - | 1 - 2 | 101 | 5050848 | 80 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK040939.1,AK052846.1,BC025007.1,NM_001170643.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
