Assay Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146207.2 | NP_666319.2 | 8 - 9 | 881 | 65 | |
| GenBank mRNA | AK144802.1 | - | 8 - 9 | 863 | 65 | |
| AK150237.1 | - | 8 - 9 | 860 | 65 | ||
| AK168215.1 | - | 8 - 9 | 863 | 65 | ||
| AK170722.1 | - | 8 - 9 | 863 | 65 | ||
| AK196574.1 | - | 1 - 2 | 51 | 65 | ||
| BC010211.1 | - | 8 - 9 | 859 | 3969515 | 65 | |
| BC024113.1 | - | 4 - 5 | 353 | 5097944 | 65 | |
| BC027006.1 | - | 4 - 5 | 401 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul4a
Genomic Map