Assay Details
| Gene Symbol: Arhgef6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152801.2 | NP_690014.2 | 9 - 10 | 1271 | 78 | |
| GenBank mRNA | AF393831.1 | - | 9 - 10 | 1271 | 78 | |
| AK006635.1 | - | 4 - 5 | 418 | 78 | ||
| AK040468.1 | - | 9 - 10 | 1093 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef6
Genomic Map