Assay Details
| Gene Symbol: Hpse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152803.4 | NP_690016.1 | 3 - 4 | 561 | 73 | |
| GenBank mRNA | AF359507.1 | - | 3 - 4 | 550 | 73 | |
| AK040471.1 | - | 3 - 4 | 562 | 73 | ||
| AK087283.1 | - | 3 - 4 | 588 | 73 | ||
| AK154628.1 | - | 3 - 4 | 562 | 73 | ||
| AY077467.1 | - | 3 - 4 | 553 | 73 | ||
| AY151051.1 | - | 3 - 4 | 472 | 73 | ||
| BC138782.1 | - | 3 - 4 | 555 | 8861804 | 73 | |
| BC138784.1 | - | 3 - 4 | 555 | 8861806 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hpse
Genomic Map