Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000002630.mm.1 |
NM_153055.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002630.mm.1 |
NM_153055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002630.mm.1 |
TC1000000386.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002627.mm.1 |
NM_153055.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002627.mm.1 |
NM_153055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002627.mm.1 |
TC1000000386.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002629.mm.1 |
NM_153055.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002629.mm.1 |
NM_153055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002629.mm.1 |
TC1000000386.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002634.mm.1 |
NM_153055.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002634.mm.1 |
NM_153055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002634.mm.1 |
TC1000000386.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002632.mm.1 |
NM_153055.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002632.mm.1 |
NM_153055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002632.mm.1 |
TC1000000386.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002631.mm.1 |
NM_153055.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002631.mm.1 |
NM_153055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002631.mm.1 |
TC1000000386.mm.1 |
MTA TC |