Assay Details
Gene Symbol: Sec63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153055.3 | NP_694695.3 | 17 - 18 | 2104 | 118 | |
GenBank mRNA | AK036629.1 | - | 17 - 18 | 1795 | 118 | |
AY024346.1 | - | 17 - 18 | 1901 | 118 | ||
AY238936.1 | - | 17 - 18 | 1874 | 118 | ||
BC019366.1 | - | 17 - 18 | 1868 | 5066908 | 118 | |
BC029774.1 | - | 9 - 10 | 1105 | 118 | ||
BC031846.1 | - | 17 - 18 | 1868 | 5066822 | 118 | |
BC059816.1 | - | 17 - 18 | 2103 | 118 | ||
BC080810.1 | - | 16 - 17 | 3405 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
