Assay Details
| Gene Symbol: Nomo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153057.4 | NP_694697.3 | 18 - 19 | 2164 | 91 | |
| GenBank mRNA | AK030584.1 | - | 18 - 19 | 2131 | 91 | |
| AK035957.1 | - | 18 - 19 | 2136 | 91 | ||
| AK080754.1 | - | 18 - 19 | 2027 | 91 | ||
| AK082495.1 | - | 18 - 19 | 2043 | 91 | ||
| AK082963.1 | - | 18 - 19 | 2023 | 91 | ||
| AK142338.1 | - | 18 - 19 | 2134 | 91 | ||
| AK150107.1 | - | 18 - 19 | 2118 | 91 | ||
| AK165643.1 | - | 18 - 19 | 2026 | 91 | ||
| AK166764.1 | - | 3 - 4 | 174 | 91 | ||
| BC022138.1 | - | 17 - 18 | 1880 | 91 | ||
| BC033923.1 | - | 7 - 8 | 748 | 5099899 | 91 | |
| BC072630.1 | - | 18 - 19 | 2110 | 6833516 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nomo1
Genomic Map