Assay Details
| Gene Symbol: Nomo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153057.4 | NP_694697.3 | 26 - 27 | 3222 | 104 | |
| GenBank mRNA | AK030584.1 | - | 26 - 27 | 3190 | 105 | |
| AK035957.1 | - | 26 - 27 | 3194 | 104 | ||
| AK080754.1 | - | 26 - 27 | 3085 | 104 | ||
| AK082495.1 | - | 26 - 27 | 3101 | 104 | ||
| AK082963.1 | - | 26 - 27 | 3081 | 104 | ||
| AK142338.1 | - | 26 - 27 | 3192 | 104 | ||
| AK165643.1 | - | 26 - 27 | 3085 | 104 | ||
| AK166764.1 | - | 11 - 12 | 1232 | 104 | ||
| BC022138.1 | - | 25 - 26 | 2938 | 104 | ||
| BC024503.1 | - | 4 - 5 | 465 | 104 | ||
| BC025133.1 | - | 9 - 10 | 1101 | 104 | ||
| BC033923.1 | - | 15 - 16 | 1806 | 5099899 | 104 | |
| BC072630.1 | - | 26 - 27 | 3168 | 6833516 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nomo1
Genomic Map