Assay Details
| Gene Symbol: Nomo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153057.4 | NP_694697.3 | 27 - 28 | 3331 | 101 | |
| GenBank mRNA | AK035957.1 | - | 27 - 28 | 3303 | 101 | |
| AK043279.1 | - | 2 - 3 | 148 | 101 | ||
| AK080754.1 | - | 27 - 28 | 3194 | 101 | ||
| AK082495.1 | - | 27 - 28 | 3210 | 101 | ||
| AK082963.1 | - | 27 - 28 | 3190 | 101 | ||
| AK142338.1 | - | 27 - 28 | 3301 | 101 | ||
| AK166764.1 | - | 12 - 13 | 1341 | 101 | ||
| AK190977.1 | - | 1 - 2 | 34 | 101 | ||
| AK200117.1 | - | 1 - 2 | 34 | 101 | ||
| AK216429.1 | - | 1 - 2 | 34 | 101 | ||
| BC022138.1 | - | 26 - 27 | 3047 | 101 | ||
| BC024503.1 | - | 5 - 6 | 574 | 101 | ||
| BC025133.1 | - | 10 - 11 | 1210 | 101 | ||
| BC033923.1 | - | 16 - 17 | 1915 | 5099899 | 101 | |
| BC072630.1 | - | 27 - 28 | 3277 | 6833516 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nomo1
Genomic Map