Assay Details
| Gene Symbol: Spns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276383.1 | NP_001263312.1 | 10 - 11 | 1227 | 89 | |
| NM_153060.3 | NP_694700.2 | 10 - 11 | 1575 | 89 | ||
| GenBank mRNA | AB441166.1 | - | 10 - 11 | 1447 | 89 | |
| AK156007.1 | - | 11 - 12 | 1221 | 89 | ||
| BC011467.1 | - | 10 - 11 | 1575 | 89 | ||
| BC025823.1 | - | 1 - 2 | 249 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
B cell homeostasis
lymphocyte homeostasis
regulation of humoral immune response
sphingosine-1-phosphate signaling pathway
sphingolipid metabolic process
transport
lipid transport
T cell homeostasis
regulation of eye pigmentation
lymph node development
transmembrane transport
bone development
lymphocyte migration
sphingolipid transporter activity
B cell homeostasis
lymphocyte homeostasis
regulation of humoral immune response
sphingosine-1-phosphate signaling pathway
sphingolipid metabolic process
transport
lipid transport
T cell homeostasis
regulation of eye pigmentation
lymph node development
transmembrane transport
bone development
lymphocyte migration
sphingolipid transporter activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018929 | BC025823.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018929 | AK156007.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018929 | BC011467.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018929 | NM_153060.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018929 | AK083908.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297247 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921391 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45908 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813723 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858928 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227964 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308816 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916088 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710633 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20931 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79282 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112375 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123565 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018929 | BC025823.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018929 | AK156007.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018929 | BC011467.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018929 | NM_153060.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018929 | AK083908.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07393604 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123565 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07393538 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112375 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07393589 | - | - |
| Whole Mouse Genome G4122A | A_51_P123745 | BC025823.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P123745 | AK156007.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P123745 | BC011467.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P123745 | NM_153060.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P123745 | AK083908.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127141 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112375 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09459043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09724786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09095812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09649083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09461658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09397316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09492307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09571008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09605566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09459038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09410681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09570998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09095805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09361759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09461637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09134268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09461642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09492300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09049423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09278011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09637649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09649079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09361762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27911731 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10388202 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388198 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388197 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388208 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388196 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388209 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388195 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388210 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388205 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388200 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388207 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388206 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388199 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451601_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10388194 | - | - |
| Mouse Genome 430 2.0 Array | 1451601_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451601_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100029182.mm.1 | NM_001276383.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100029182.mm.1 | NM_153060.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100029182.mm.1 | TC1100003146.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451601_PM_a_at | - | - |
| Mouse Expression Set 430 | 1451601_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451601_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451601_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451601_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451601_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 905012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6620121 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6620121 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006620121 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2660293 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2660293 | - | - |
| mouse-6 v1.1 (Illumina) | scl39968.13_19-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6620121 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39968.13_19-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620121 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2660293 | - | - |

Spns2
Genomic Map