Assay Details
| Gene Symbol: Ddx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153065.3 | NP_694705.2 | 5 - 6 | 528 | 99 | |
| GenBank mRNA | AK134274.1 | - | 5 - 6 | 529 | 99 | |
| AK138442.1 | - | 5 - 6 | 552 | 99 | ||
| AK146469.1 | - | 5 - 6 | 552 | 99 | ||
| AK182136.1 | - | 4 - 5 | 354 | 99 | ||
| BC011321.1 | - | 4 - 5 | 405 | 4007058 | 99 | |
| BC024730.1 | - | 5 - 6 | 528 | 99 | ||
| BC024981.1 | - | 5 - 6 | 528 | 99 | ||
| BC033398.1 | - | 5 - 6 | 528 | 99 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC011321.1 transcripts.
Set Membership: |
|

Ddx27
Genomic Map