Assay Details
| Gene Symbol: Prss42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153099.1 | NP_694739.1 | 4 - 5 | 805 | 99 | |
| GenBank mRNA | AB052292.1 | - | 4 - 5 | 805 | 99 | |
| AK133402.1 | - | 4 - 5 | 670 | 99 | ||
| BC060984.1 | - | 4 - 5 | 824 | 6774705 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242487 | AK133402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242487 | BC060984.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242487 | NM_153099.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242487 | AB052292.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242487 | AK133402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242487 | BC060984.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242487 | NM_153099.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242487 | AB052292.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737157 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155804 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128076 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242487 | AK133402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242487 | BC060984.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242487 | NM_153099.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242487 | AB052292.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242487 | AK133402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242487 | BC060984.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242487 | NM_153099.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242487 | AB052292.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825929 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128076 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06926138 | - | - |
| Whole Mouse Genome G4122A | A_51_P242487 | AK133402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P242487 | BC060984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P242487 | NM_153099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P242487 | AB052292.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P242487 | AK133402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P242487 | BC060984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P242487 | NM_153099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P242487 | AB052292.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P441589 | AK133402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P441589 | BC060984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P441589 | NM_153099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P441589 | AB052292.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130775 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128076 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07994266 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07994282 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07994268 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07782365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07624433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08035597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07750544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07784722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07526848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07485202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07981837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07981839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07485197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07544381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07624434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07739671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08127134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07782365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07485196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07526847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08124433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07944024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07994266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07994280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26825923 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10589615 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589613 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589611 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589612 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589614 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425169_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10589610 | - | - |
| Mouse Genome 430 2.0 Array | 1425169_at | - | - |
| Mouse Genome 430 2.0 Array | 1425169_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235628_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235628_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900012296.mm.1 | NM_153099.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012296.mm.1 | NM_153099.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012296.mm.1 | TC0900001469.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425169_PM_at | - | - |
| Mouse Expression Set 430 | 1425169_at | - | - |
| Murine Genome U74v2 Set | 167400_i_at | - | - |
| Murine Genome U74v2 Set | 167400_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425169_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425169_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425169_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425169_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 808197 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 7040500 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007040500 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224452 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224452 | - | - |
| mouse-6 v1.1 (Illumina) | scl36422.5.1_67-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7040500 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36422.5.1_67-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040500 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2878791 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2878791 | - | - |

Prss42
Genomic Map