Assay Details
Gene Symbol: Cpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153107.2 | NP_694747.2 | 4 - 5 | 813 | 85 | |
GenBank mRNA | AF356844.1 | - | 4 - 5 | 722 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
