Assay Details
Gene Symbol: Unc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153131.3 | NP_694771.1 | 4 - 5 | 768 | 92 | |
GenBank mRNA | AJ487852.1 | - | 4 - 5 | 769 | 92 | |
AK122575.1 | - | 4 - 5 | 536 | 92 | ||
BC058084.1 | - | 4 - 5 | 789 | 6813463 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
