Assay Details
| Gene Symbol: Traf3ip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153137.4 | NP_694777.3 | 8 - 9 | 1122 | 61 | |
| GenBank mRNA | AK031400.1 | - | 8 - 9 | 1062 | 61 | |
| AK036608.1 | - | 6 - 7 | 1252 | 61 | ||
| AK161211.1 | - | 5 - 6 | 732 | 61 | ||
| AY383616.1 | - | 6 - 7 | 706 | 61 | ||
| BC036561.1 | - | 8 - 9 | 930 | 61 | ||
| BC137605.1 | - | 6 - 7 | 733 | 8860624 | 61 | |
| BC171933.1 | - | 6 - 7 | 733 | 9054217 | 61 | |
More Information
Set Membership: |
|

Traf3ip3
Genomic Map