Assay Details
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162868.1 | NP_001156340.1 | 13 - 14 | 3028 | 70 | |
NM_001162869.1 | NP_001156341.1 | 14 - 15 | 3163 | 70 | ||
NM_153140.2 | NP_694780.1 | 11 - 12 | 1419 | 70 | ||
GenBank mRNA | AB093257.1 | - | 14 - 15 | 2604 | 70 | |
AK085516.1 | - | 11 | 1455 | 70 | ||
AK128985.1 | - | 11 - 12 | 1419 | 70 | ||
BC023364.1 | - | 11 - 12 | 1410 | 70 | ||
BC037132.1 | - | 11 - 12 | 1459 | 70 | ||
CB840427.1 | - | 2 - 3 | 314 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082156 | CF747684.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082156 | BC037132.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082156 | BC023364.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082156 | AK128985.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082156 | AK083658.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784929 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 965947 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713450 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208715 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71445 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346040 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130285 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947174 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198963 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422235 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777693 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681418 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777453 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851449 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823827 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 49180 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176286 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869509 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786527 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248074 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179143 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159293 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85759 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964351 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338627 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53871 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549275 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238442 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75550 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734313 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485492 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612576 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963613 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737975 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139873 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652330 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771518 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891898 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243453 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862584 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727145 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482824 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123599 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46855 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87328 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112447 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50314 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57914 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38555 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31607 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113273 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160341 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154017 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122956 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082156 | CF747684.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082156 | BC037132.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082156 | BC023364.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082156 | AK128985.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082156 | AK083658.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03296434 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08744284 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122956 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08744334 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154017 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03296463 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08744279 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03296499 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03296529 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08744369 | - | - |
Whole Mouse Genome G4122A | A_51_P431491 | CF747684.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431491 | BC037132.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431491 | BC023364.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431491 | AK128985.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431491 | AK083658.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154017 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08744369 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122956 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08744334 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13483065 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107885 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13277141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107877 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108085 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108092 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107890 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107886 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107869 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13373263 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13123405 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108133 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107682 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107698 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108107 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13152113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13498345 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107915 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108138 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13413417 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108093 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107977 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108117 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107706 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13386173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107895 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13498343 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107868 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13244171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13277137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13413165 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107701 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13139050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13244173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13154751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13555740 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107874 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107912 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13373281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107687 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107979 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107904 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108080 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13121829 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107703 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108089 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108095 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107906 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107888 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108086 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107694 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13373260 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107883 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13413164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13413147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107693 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107872 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108087 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107881 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13277140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13373262 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108090 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107884 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107893 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31107710 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13167926 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108135 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31108114 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10449220 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449224 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449217 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449223 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449209 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449219 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449214 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449208 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449211 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449215 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449210 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449221 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449212 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449222 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449218 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449216 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10449213 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438022_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1457864_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10449207 | - | - |
Mouse Genome 430 2.0 Array | 1438022_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457864_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457864_at | - | - |
Mouse Genome 430 2.0 Array | 1438022_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 215445_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 215445_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700015637.mm.1 | NM_001162868.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700015637.mm.1 | NM_001162868.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700015637.mm.1 | NM_001162869.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700015637.mm.1 | NM_001162869.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700015637.mm.1 | NM_153140.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700015637.mm.1 | NM_153140.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700015637.mm.1 | TC1700001686.mm.1 | MTA TC |
Mouse Expression Set 430 | 1457864_PM_at | - | - |
Mouse Expression Set 430 | 1438022_PM_at | - | - |
Mouse Expression Set 430 | 1438022_at | - | - |
Mouse Expression Set 430 | 1457864_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438022_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457864_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438022_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457864_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2510070 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2510070 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002510070 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2638520 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2638520 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1248291 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1227748 | - | - |
mouse-6 v1.1 (Illumina) | scl50156.18_539-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2510070 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl50156.18_539-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2510070 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2638520 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2638520 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50156.18_539-S | - | - |
