Assay Details
Gene Symbol: Acat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153151.3 | NP_694791.2 | 8 - 9 | 1123 | 95 | |
GenBank mRNA | AF356876.1 | - | 8 - 9 | 95 | ||
AK143884.1 | - | 8 - 9 | 1083 | 95 | ||
AK149441.1 | - | 8 - 9 | 1123 | 95 | ||
BC049873.1 | - | 8 - 9 | 1028 | 5054536 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101407 | BC049873.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101407 | AF356876.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101407 | AK143884.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101407 | AK149441.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101407 | NM_153151.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162798 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952027 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436102 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795896 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111542 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251534 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39525 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129985 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164167 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171583 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101407 | BC049873.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101407 | AF356876.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101407 | AK143884.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101407 | AK149441.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101407 | NM_153151.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055281 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055288 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055287 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055286 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055289 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055304 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055302 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055284 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055300 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055278 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055283 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055279 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055301 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164167 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171583 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08720797 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08720806 | - | - |
Whole Mouse Genome G4122A | A_51_P139745 | M35797.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P11959 | BC049873.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P11959 | AF356876.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P11959 | AK143884.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P11959 | AK149441.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P11959 | NM_153151.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101407 | BC049873.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101407 | AF356876.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101407 | AK143884.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101407 | AK149441.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101407 | NM_153151.3 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171583 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164167 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13522782 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13317960 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13317962 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13522787 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13317959 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13317958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055427 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055270 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055283 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055315 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055272 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055305 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055308 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055284 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13510109 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055279 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13317962 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055276 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055286 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055306 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31055310 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10447887 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10447890 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10447889 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10447886 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10447888 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10447885 | - | - |
Mouse Genome 430 2.0 Array | 1425195_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1425195_a_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700013030.mm.1 | NM_153151.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013030.mm.1 | NM_153151.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013030.mm.1 | TC1700001458.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013032.mm.1 | NM_153151.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013032.mm.1 | NM_153151.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013032.mm.1 | TC1700001458.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013033.mm.1 | NM_153151.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013033.mm.1 | NM_153151.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013033.mm.1 | TC1700001458.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013035.mm.1 | NM_153151.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013035.mm.1 | NM_153151.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013035.mm.1 | TC1700001458.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013037.mm.1 | NM_153151.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013037.mm.1 | NM_153151.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013037.mm.1 | TC1700001458.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013029.mm.1 | NM_153151.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013029.mm.1 | NM_153151.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013029.mm.1 | TC1700001458.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013034.mm.1 | NM_153151.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013034.mm.1 | NM_153151.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013034.mm.1 | TC1700001458.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013031.mm.1 | NM_153151.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013031.mm.1 | NM_153151.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013031.mm.1 | TC1700001458.mm.1 | MTA TC |
Mouse Expression Set 430 | 1425195_a_at | - | - |
Mouse Expression Set 430 | 1425195_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1425195_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1425195_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 730737 | - | - |
ABI Mouse Genome Survey Microarray | 749204 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 610097 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 610097 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1940592 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2630408 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002630408 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000610097 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001940592 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252807 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2748150 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1212761 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1252807 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1212761 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2748150 | - | - |
mouse-6 v1.1 (Illumina) | scl0224530.9_22-S | - | - |
mouse-6 v1.1 (Illumina) | scl00224530.2_232-S | - | - |
mouse-6 v1.1 (Illumina) | scl00224530.2_63-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2630408 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 610097 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1940592 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0224530.9_22-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00224530.2_232-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00224530.2_63-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630408 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940592 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 610097 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1216631 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216631 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2748150 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00224530.2_232-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00224530.2_63-S | - | - |
