Assay Details
| Gene Symbol: Prss45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153172.1 | NP_694812.1 | 5 - 6 | 762 | 86 | |
| GenBank mRNA | AF392062.1 | - | 5 - 6 | 762 | 86 | |
| AK077121.1 | - | 5 - 6 | 760 | 86 | ||
| BC107230.1 | - | 4 - 5 | 735 | 40052042 | 86 | |
| BC107231.1 | - | 4 - 5 | 735 | 40052043 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26826133 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26826132 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26826131 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505914 | BC107231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505914 | NM_153172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505914 | AF392062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505914 | AK077121.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505914 | BC107230.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505914 | BC107231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505914 | NM_153172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505914 | AF392062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505914 | AK077121.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505914 | BC107230.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367533 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 679830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11868 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114035 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180284 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505914 | BC107231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505914 | NM_153172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505914 | AF392062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505914 | AK077121.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505914 | BC107230.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505914 | BC107231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505914 | NM_153172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505914 | AF392062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505914 | AK077121.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505914 | BC107230.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826164 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06926248 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180284 | - | - |
| Whole Mouse Genome G4122A | A_51_P505914 | BC107231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505914 | NM_153172.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505914 | AF392062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505914 | AK077121.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505914 | BC107230.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505914 | BC107231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505914 | NM_153172.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505914 | AF392062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505914 | AK077121.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505914 | BC107230.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104732 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180284 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07621513 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07621515 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07621505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08061538 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07621504 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07778977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26826133 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26826131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26826132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07503008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07923899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07836594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07621505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07863430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07829282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07778977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07923891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08143795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07863436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07836596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07923886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07621509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26826119 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10589634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589635 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589638 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456452_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10589632 | - | - |
| Mouse Genome 430 2.0 Array | 1456452_at | - | - |
| Mouse Genome 430 2.0 Array | 1456452_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 260408_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 260408_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900012324.mm.1 | NM_153172.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012324.mm.1 | NM_153172.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012324.mm.1 | TC0900001472.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900012325.mm.1 | NM_153172.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012325.mm.1 | NM_153172.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012325.mm.1 | TC0900001472.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456452_at | - | - |
| Mouse Expression Set 430 | 1456452_PM_at | - | - |
| Murine Genome U74v2 Set | 167805_PM_i_at | - | - |
| Murine Genome U74v2 Set | 167805_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456452_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456452_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 770018 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2900706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2900706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245069 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245069 | - | - |
| mouse-6 v1.1 (Illumina) | scl36419.6.1_168-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36419.6.1_168-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245069 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245069 | - | - |

Prss45
Genomic Map