Assay Details
| Gene Symbol: Hist1h4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007642.1 | - | 1 | 432 | 83 | |
| BC028550.1 | - | 1 | 377 | 1398106 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28881996 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28881999 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130646 | AA028777.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130646 | AK007642.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130646 | BC087952.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130646 | NM_153173.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130646 | AA028777.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130646 | AK007642.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130646 | BC087952.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130646 | NM_153173.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881957 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02646062 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130646 | AA028777.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130646 | AK007642.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130646 | BC087952.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130646 | NM_153173.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02646062 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10960792 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11103145 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10960801 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10960788 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28881999 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28881996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10816526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10889690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10546567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10586514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10816534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10816536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881931 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404025 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408075 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403958 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404068 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428014_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404024 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408074 | - | - |
| Mouse Genome 430 2.0 Array | 1422948_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1428014_at | - | - |
| Mouse Genome 430 2.0 Array | 1431333_at | - | - |
| Mouse Genome 430 2.0 Array | 1431333_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428014_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422948_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69386_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69386_at | - | - |
| Mouse Expression Set 430 | 1428014_at | - | - |
| Mouse Expression Set 430 | 1422947_at | - | - |
| Mouse Expression Set 430 | 1424854_PM_at | - | - |
| Mouse Expression Set 430 | 1428014_PM_at | - | - |
| Mouse Expression Set 430 | 1422948_PM_s_at | - | - |
| Mouse Expression Set 430 | 1424854_at | - | - |
| Mouse Expression Set 430 | 1422947_PM_at | - | - |
| Mouse Expression Set 430 | 1422948_s_at | - | - |
| Mouse Expression Set 430 | 1431658_at | - | - |
| Mouse Expression Set 430 | 1431615_PM_at | - | - |
| Mouse Expression Set 430 | 1431658_PM_at | - | - |
| Mouse Expression Set 430 | 1431615_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422948_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422948_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422947_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428014_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424854_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422947_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428014_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424854_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104480180 | - | - |
| MouseWG-6 v2 Expression BeadChip | 103360632 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103360632 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4060446 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104480180 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060446 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2664129 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2439060 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664129 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2525554 | - | - |
| mouse-6 v1.1 (Illumina) | scl44214.2.4_53-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0069386.2_29-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_23943921-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103360632 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104480180 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060446 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0069386.2_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103360632 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104480180 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060446 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2664129 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2439060 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2664129 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0069386.2_29-S | - | - |

Hist1h4h
Genomic Map