Assay Details
Gene Symbol: Tubgcp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290824.1 | NP_001277753.1 | 3 - 4 | 562 | 102 | |
NM_153387.3 | NP_700436.1 | 3 - 4 | 562 | 102 | ||
NR_077245.2 | 3 - 4 | 562 | 102 | |||
NR_110985.1 | 3 - 4 | 562 | 102 | |||
GenBank mRNA | AK016563.1 | - | 3 - 4 | 605 | 102 | |
AK045305.1 | - | 3 - 4 | 455 | 102 | ||
AK046132.1 | - | 3 - 4 | 492 | 102 | ||
AK050250.1 | - | 3 - 4 | 563 | 102 | ||
AK051517.1 | - | 3 - 4 | 463 | 102 | ||
AK152984.1 | - | 3 - 4 | 521 | 102 | ||
AK153171.1 | - | 3 - 4 | 521 | 102 | ||
BC029106.1 | - | 3 - 4 | 526 | 3711922 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
