Assay Details
Gene Symbol: Col23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153393.2 | NP_700442.2 | 17 - 18 | 1201 | 135 | |
GenBank mRNA | AF410792.1 | - | 17 - 18 | 1201 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
