Assay Details
| Gene Symbol: Ints9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253731.1 | NP_001240660.1 | 14 - 15 | 1915 | 84 | |
| NM_153414.4 | NP_700463.2 | 14 - 15 | 1883 | 84 | ||
| GenBank mRNA | AK077634.1 | - | 14 - 15 | 1884 | 84 | |
| AK136992.1 | - | 14 - 15 | 1610 | 84 | ||
| AK150195.1 | - | 14 - 15 | 1610 | 84 | ||
| BC028953.1 | - | 13 - 14 | 1573 | 5362016 | 84 | |
| BC055700.1 | - | 14 - 15 | 1582 | 6808816 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ints9
Genomic Map