Assay Details
Gene Symbol: Ints9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253731.1 | NP_001240660.1 | 15 - 16 | 2004 | 87 | |
NM_153414.4 | NP_700463.2 | 15 - 16 | 1972 | 87 | ||
GenBank mRNA | AK077634.1 | - | 15 - 16 | 1973 | 87 | |
AK136992.1 | - | 15 - 16 | 1699 | 87 | ||
AK150195.1 | - | 15 - 16 | 1699 | 87 | ||
BC028953.1 | - | 14 - 15 | 1662 | 5362016 | 87 | |
BC055700.1 | - | 15 - 16 | 1671 | 6808816 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
