Assay Details
| Gene Symbol: Aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153416.2 | NP_700465.2 | 3 - 4 | 448 | 91 | |
| GenBank mRNA | AK034591.1 | - | 3 - 4 | 343 | 91 | |
| AK079849.1 | - | 3 - 4 | 385 | 91 | ||
| AK083401.1 | - | 3 - 4 | 361 | 91 | ||
| AK083537.1 | - | 3 - 4 | 369 | 91 | ||
| AK087134.1 | - | 3 - 4 | 423 | 92 | ||
| AK088247.1 | - | 3 - 4 | 365 | 91 | ||
| AK134406.1 | - | 3 - 4 | 339 | 91 | ||
| AK165243.1 | - | 3 - 4 | 363 | 91 | ||
| AK166118.1 | - | 3 - 4 | 363 | 91 | ||
| AK169523.1 | - | 3 - 4 | 400 | 91 | ||
| AK189195.1 | - | 2 - 3 | 138 | 92 | ||
| BC018191.1 | - | 3 - 4 | 349 | 3994248 | 91 | |
| BC025501.1 | - | 3 - 4 | 395 | 5321013 | 91 | |
| BC026604.1 | - | 2 - 3 | 617 | 91 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK087134.1 transcripts.
Set Membership: |
|

Aaas
Genomic Map