Assay Details
| Gene Symbol: Aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153416.2 | NP_700465.2 | 4 - 5 | 537 | 63 | |
| GenBank mRNA | AK034591.1 | - | 4 - 5 | 432 | 63 | |
| AK079849.1 | - | 4 - 5 | 474 | 63 | ||
| AK083401.1 | - | 4 - 5 | 450 | 63 | ||
| AK083537.1 | - | 4 - 5 | 458 | 63 | ||
| AK087134.1 | - | 4 - 5 | 513 | 63 | ||
| AK088247.1 | - | 4 - 5 | 454 | 63 | ||
| AK134406.1 | - | 4 - 5 | 428 | 63 | ||
| AK165243.1 | - | 4 - 5 | 452 | 63 | ||
| AK166118.1 | - | 4 - 5 | 452 | 63 | ||
| AK169523.1 | - | 4 - 5 | 489 | 63 | ||
| AK189195.1 | - | 3 - 4 | 228 | 63 | ||
| BC018191.1 | - | 4 - 5 | 438 | 3994248 | 63 | |
| BC025501.1 | - | 4 - 5 | 484 | 5321013 | 63 | |
| BC026604.1 | - | 3 - 4 | 706 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK087134.1 transcripts.
Set Membership: |
|

Aaas
Genomic Map