Assay Details
| Gene Symbol: Opalin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153520.1 | NP_705740.1 | 2 - 3 | 111 | 114 | |
| GenBank mRNA | AK140031.1 | - | 112 | 114 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK140031.1,NM_153520.1 transcripts.
Set Membership: |
|

Opalin
Genomic Map