Assay Details
| Gene Symbol: Lrrc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153545.2 | NP_705773.2 | 1 - 2 | 104 | 95 | |
| GenBank mRNA | AK081566.1 | - | 1 - 2 | 107 | 95 | |
| AK153361.1 | - | 1 - 2 | 105 | 95 | ||
| BC023296.1 | - | 1 - 2 | 84 | 3662961 | 95 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK153361.1,BC023296.1,NM_153545.2 transcripts.
Set Membership: |
|

Lrrc45
Genomic Map