Assay Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098114.1 | - | 5 - 6 | 1113 | 100 | |
| BC023704.1 | - | 6 - 7 | 1035 | 5345701 | 100 | |
| BC023804.1 | - | 6 - 7 | 1148 | 5352941 | 100 | |
| BC078641.1 | - | 5 - 6 | 2006 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf8
Genomic Map