Assay Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153555.2 | NP_705783.1 | 10 - 11 | 1535 | 149 | |
| GenBank mRNA | AK035701.1 | - | 10 - 11 | 1434 | 149 | |
| AK098114.1 | - | 10 - 11 | 1554 | 149 | ||
| AK146825.1 | - | 10 - 11 | 1462 | 149 | ||
| AK162622.1 | - | 8 - 9 | 2483 | 149 | ||
| AK167059.1 | - | 10 - 11 | 1411 | 149 | ||
| AK194034.1 | - | 4 - 5 | 149 | |||
| BC023704.1 | - | 10 - 11 | 1418 | 5345701 | 149 | |
| BC023804.1 | - | 11 - 12 | 1589 | 5352941 | 149 | |
| BC078641.1 | - | 10 - 11 | 2447 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf8
Genomic Map