Assay Details
| Gene Symbol: Obp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153558.1 | NP_705786.1 | 6 - 7 | 553 | 95 | |
| GenBank mRNA | AY360148.1 | - | 6 - 7 | 589 | 95 | |
| BC027556.1 | - | 6 - 7 | 553 | 1434737 | 95 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135153 | AY360148.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135153 | BC027556.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135153 | NM_153558.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 949097 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146761 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109006 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104515 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135153 | AY360148.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135153 | BC027556.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135153 | NM_153558.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073867 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146761 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104515 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109006 | - | - |
| Whole Mouse Genome G4122A | A_51_P479698 | AY360148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479698 | BC027556.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479698 | NM_153558.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104378 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126795 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109006 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104515 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146761 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01949800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01949839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01554685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01949830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01949804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02003035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01819426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01949829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01554687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02267913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01949841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073850 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10470176 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470177 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470181 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470178 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428022_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10470175 | - | - |
| Mouse Genome 430 2.0 Array | 1428022_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428022_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 227627_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 227627_PM_at | - | - |
| Mouse Expression Set 430 | 1428022_at | - | - |
| Mouse Expression Set 430 | 1428022_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428022_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428022_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428022_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428022_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 365758 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6110435 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110435 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2695360 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2695360 | - | - |
| mouse-6 v1.1 (Illumina) | scl21166.7.1_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110435 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21166.7.1_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110435 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2695360 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2695360 | - | - |

Obp2a
Genomic Map