Assay Details
| Gene Symbol: Ino80e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153580.1 | NP_705808.1 | 1 - 2 | 219 | 110 | |
| GenBank mRNA | BC026475.1 | - | 1 - 2 | 219 | 4489168 | 110 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25502648 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25502649 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999434 | BC026475.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999434 | NM_153580.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133217 | BC055928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133217 | BC025527.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999439 | AK135531.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999439 | BC025527.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221866 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677125 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 464204 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161463 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102268 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125083 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143453 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133217 | BC055928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133217 | BC025527.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999439 | AK135531.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999439 | BC025527.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999434 | BC026475.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999434 | NM_153580.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502619 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102268 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143453 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161463 | - | - |
| Whole Mouse Genome G4122A | A_52_P111562 | BC055928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P111562 | AK135531.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P111562 | BC019595.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P111562 | BC025527.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P111562 | BC026475.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102679 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143453 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102268 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161463 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06333492 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06333490 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06333502 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06333482 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25502649 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25502648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06664430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06245461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06190591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06211286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06132842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06480849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06041916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502624 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568074 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568076 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568073 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568075 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568077 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451347_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424429_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424428_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10557508 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568065 | - | - |
| Mouse Genome 430 2.0 Array | 1424428_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424428_at | - | - |
| Mouse Genome 430 2.0 Array | 1424429_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1451347_at | - | - |
| Mouse Genome 430 2.0 Array | 1451347_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424429_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233875_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233875_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700036008.mm.1 | NM_153580.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036008.mm.1 | NM_153580.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036008.mm.1 | TC0700004301.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036005.mm.1 | NM_153580.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036005.mm.1 | NM_153580.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036005.mm.1 | TC0700004301.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451347_at | - | - |
| Mouse Expression Set 430 | 1424428_at | - | - |
| Mouse Expression Set 430 | 1424429_s_at | - | - |
| Mouse Expression Set 430 | 1451347_PM_at | - | - |
| Mouse Expression Set 430 | 1424428_PM_at | - | - |
| Mouse Expression Set 430 | 1424429_PM_s_at | - | - |
| Murine Genome U74v2 Set | 99949_PM_at | - | - |
| Murine Genome U74v2 Set | 99949_at | - | - |
| Murine Genome U74v2 Set | 171391_at | - | - |
| Murine Genome U74v2 Set | 171391_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451347_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424428_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424429_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424428_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424429_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451347_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424429_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451347_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424428_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424429_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424428_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451347_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101980519 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5270253 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3130541 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101980519 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5270253 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270253 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003130541 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244842 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244842 | - | - |
| mouse-6 v1.1 (Illumina) | scl00233875.2_247-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00085.1_46-S | - | - |
| mouse-6 v1.1 (Illumina) | scl30668.7_258-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101980519 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270253 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3130541 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00085.1_46-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00233875.2_247-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101980519 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270253 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130541 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1244842 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2876560 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244842 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2876560 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00233875.2_247-S | - | - |

Ino80e
Genomic Map