Assay Details
| Gene Symbol: Klre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153590.3 | NP_705818.3 | 6 - 7 | 668 | 116 | |
| GenBank mRNA | AF306663.1 | - | 6 - 7 | 668 | 116 | |
| BX518942.1 | - | 2 - 3 | 246 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
stimulatory C-type lectin receptor signaling pathway
negative regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
natural killer cell activation
positive regulation of interferon-gamma production
positive regulation of natural killer cell mediated cytotoxicity
transmembrane signaling receptor activity
protein phosphatase binding
protein homodimerization activity
protein dimerization activity
stimulatory C-type lectin receptor signaling pathway
negative regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
natural killer cell activation
positive regulation of interferon-gamma production
positive regulation of natural killer cell mediated cytotoxicity
transmembrane signaling receptor activity
protein phosphatase binding
protein homodimerization activity
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071716 | AK041224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071716 | AY100458.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071716 | AF306663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071716 | BX518942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071716 | NM_153590.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274194 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697223 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47653 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140159 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071716 | AK041224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071716 | AY100458.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071716 | AF306663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071716 | BX518942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071716 | NM_153590.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844292 | - | - |
| Whole Mouse Genome G4122A | A_52_P592222 | AY100458.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592222 | AF306663.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592222 | BG145575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592222 | NM_153590.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P383544 | AK041224.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106935 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05853890 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05499000 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05853889 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05853884 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05853892 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05499001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05853890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05769777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05338846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05769778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05357353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05762560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05726224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844347 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10542210 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542213 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542209 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542206 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542212 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542208 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542207 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458642_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10542205 | - | - |
| Mouse Genome 430 2.0 Array | 1458642_at | - | - |
| Mouse Genome 430 2.0 Array | 1458642_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243655_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243655_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600012537.mm.1 | TC0600001537.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012537.mm.1 | NM_153590.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012537.mm.1 | NM_153590.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012538.mm.1 | TC0600001537.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012538.mm.1 | NM_153590.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012538.mm.1 | NM_153590.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012535.mm.1 | TC0600001537.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012535.mm.1 | NM_153590.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012535.mm.1 | NM_153590.3 | Refseq |
| Mouse Expression Set 430 | 1458642_PM_at | - | - |
| Mouse Expression Set 430 | 1458642_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458642_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458642_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 631030 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4850500 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850500 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239346 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239346 | - | - |
| mouse-6 v1.1 (Illumina) | scl29461.5.2_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850500 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29461.5.2_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850500 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239346 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239346 | - | - |

Klre1
Genomic Map