Assay Details
| Gene Symbol: Pcmtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291211.1 | NP_001278140.1 | 3 - 4 | 648 | 91 | |
| NM_153594.3 | NP_705822.1 | 3 - 4 | 648 | 91 | ||
| GenBank mRNA | AB182640.1 | - | 3 - 4 | 589 | 91 | |
| AK031335.1 | - | 3 - 4 | 603 | 91 | ||
| AK088800.1 | - | 3 - 4 | 587 | 91 | ||
| AK146080.1 | - | 3 - 4 | 595 | 91 | ||
| BC031360.1 | - | 2 - 3 | 465 | 5359242 | 91 | |
| BC040385.1 | - | 3 - 4 | 599 | 3499499 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcmtd2
Genomic Map