Assay Details
| Gene Symbol: Trpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153597.2 | NP_705825.2 | 3 - 4 | 644 | 60 | |
| GenBank mRNA | AK048558.1 | - | 793 | 60 | ||
| AK090183.1 | - | 2 - 3 | 1775 | 60 | ||
| BC027575.1 | - | 3 - 4 | 442 | 1364225 | 60 | |
More Information
Set Membership: |
|

Trpt1
Genomic Map