Assay Details
Gene Symbol: Mill2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153760.2 | NP_715641.1 | 2 - 3 | 387 | 98 | |
NM_153761.3 | NP_715642.2 | 3 - 4 | 432 | 98 | ||
GenBank mRNA | AB086266.1 | - | 2 - 3 | 387 | 98 | |
AB086267.1 | - | 3 - 4 | 432 | 98 | ||
BC137748.1 | - | 2 - 3 | 385 | 8860767 | 98 | |
BC144886.1 | - | 3 - 4 | 430 | 9053434 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune response-activating cell surface receptor signaling pathway
response to oxidative stress
response to heat
antigen processing and presentation
response to retinoic acid
gamma-delta T cell activation
negative regulation of defense response to virus by host
antigen binding
protein binding
natural killer cell lectin-like receptor binding
immune response-activating cell surface receptor signaling pathway
response to oxidative stress
response to heat
antigen processing and presentation
response to retinoic acid
gamma-delta T cell activation
negative regulation of defense response to virus by host
antigen binding
protein binding
natural killer cell lectin-like receptor binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231549 | AK137167.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231549 | AB086266.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231549 | AB086267.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231549 | BC137748.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231549 | BC144886.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231549 | AK137167.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231549 | AB086266.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231549 | AB086267.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231549 | BC137748.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231549 | BC144886.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809485 | AK046421.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809485 | AK046609.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809485 | NM_080454.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809485 | NM_175452.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945501 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723108 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91513 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819719 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7096 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942606 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259892 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 418081 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174118 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65607 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167175 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156960 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231549 | AK137167.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231549 | AB086266.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231549 | AB086267.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231549 | BC137748.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231549 | BC144886.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231549 | AK137167.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231549 | AB086266.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231549 | AB086267.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231549 | BC137748.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231549 | BC144886.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990123 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990119 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167175 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156960 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01483046 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06133474 | - | - |
Whole Mouse Genome G4122A | A_51_P231549 | AK137167.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P231549 | AB086266.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P231549 | AB086267.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P231549 | BC137748.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P231549 | BC144886.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P231549 | AK137167.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P231549 | AB086266.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P231549 | AB086267.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P231549 | BC137748.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P231549 | BC144886.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100949 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156960 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167175 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06082703 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06152215 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05980801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990134 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990128 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06376716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06321516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06326921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05980801 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06082701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05959943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06001952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05959940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05950333 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990134 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05950331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24990166 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10550502 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10550504 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10550505 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10550508 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10550506 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10550503 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10550501 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700001979.mm.1 | TC0700000272.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700001979.mm.1 | NM_153760.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001979.mm.1 | NM_153760.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001979.mm.1 | NM_153761.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001979.mm.1 | NM_153761.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001980.mm.1 | TC0700000272.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700001980.mm.1 | NM_153760.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001980.mm.1 | NM_153760.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001980.mm.1 | NM_153761.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001980.mm.1 | NM_153761.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001981.mm.1 | TC0700000272.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700001981.mm.1 | NM_153760.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001981.mm.1 | NM_153760.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001981.mm.1 | NM_153761.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700001981.mm.1 | NM_153761.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6350279 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006350279 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2747176 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2747176 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2542237 | - | - |
mouse-6 v1.1 (Illumina) | scl00243864.1_138-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6350279 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00243864.1_138-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350279 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2747176 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3113845 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2747176 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3113845 | - | - |
