Assay Details
| Gene Symbol: Glb1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153803.1 | NP_722498.1 | 6 - 7 | 646 | 104 | |
| GenBank mRNA | AK033400.1 | - | 2 - 3 | 229 | 104 | |
| AK136711.1 | - | 6 - 7 | 680 | 104 | ||
| AK143057.1 | - | 5 - 6 | 577 | 104 | ||
| BC038479.1 | - | 6 - 7 | 646 | 4503030 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glb1l2
Genomic Map