Assay Details
Gene Symbol: Glb1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153803.1 | NP_722498.1 | 9 - 10 | 895 | 72 | |
GenBank mRNA | AK033400.1 | - | 5 - 6 | 478 | 72 | |
AK136711.1 | - | 9 - 10 | 929 | 72 | ||
AK143057.1 | - | 8 - 9 | 826 | 72 | ||
BC038479.1 | - | 9 - 10 | 895 | 4503030 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
