Assay Details
| Gene Symbol: Arhgap15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153820.3 | NP_722542.2 | 4 - 5 | 382 | 124 | |
| GenBank mRNA | AK018058.1 | - | 4 - 5 | 351 | 124 | |
| AK031022.1 | - | 4 - 5 | 338 | 124 | ||
| AK036520.1 | - | 4 - 5 | 383 | 124 | ||
| AK041323.1 | - | 4 - 5 | 375 | 124 | ||
| AK170537.1 | - | 4 - 5 | 380 | 124 | ||
| BC024887.1 | - | 4 - 5 | 336 | 3156334 | 124 | |
| BC034881.1 | - | 4 - 5 | 355 | 3468639 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap15
Genomic Map