Assay Details
| Gene Symbol: Kcnn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163510.1 | NP_001156982.1 | 5 - 6 | 1093 | 102 | |
| NM_008433.4 | NP_032459.3 | 5 - 6 | 1093 | 102 | ||
| GenBank mRNA | AB128930.1 | - | 3 - 4 | 856 | 102 | |
| AF042487.1 | - | 5 - 6 | 959 | 102 | ||
| AF072884.1 | - | 5 - 6 | 919 | 102 | ||
| AK010943.1 | - | 5 - 6 | 1094 | 102 | ||
| AK085752.1 | - | 5 - 6 | 910 | 102 | ||
| AK155910.1 | - | 5 - 6 | 1058 | 102 | ||
| BC010274.1 | - | 5 - 6 | 1042 | 3589411 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnn4
Genomic Map