Assay Details
| Gene Symbol: Prkdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011159.2 | NP_035289.2 | 78 - 79 | 11196 | 58 | |
| GenBank mRNA | AB007544.1 | - | 78 - 79 | 11173 | 58 | |
| AB011543.1 | - | 78 - 79 | 11185 | 58 | ||
| AK088981.1 | - | 3 - 4 | 323 | 58 | ||
| AK137732.1 | - | 5 - 6 | 631 | 58 | ||
| D83786.1 | - | 3 - 4 | 331 | 58 | ||
| D87521.1 | - | 78 - 79 | 11173 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkdc
Genomic Map