Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500018826.mm.1 |
TC0500002073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018826.mm.1 |
NM_013569.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018826.mm.1 |
NM_013569.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018831.mm.1 |
TC0500002073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018831.mm.1 |
NM_013569.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018831.mm.1 |
NM_013569.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018829.mm.1 |
TC0500002073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018829.mm.1 |
NM_013569.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018829.mm.1 |
NM_013569.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018830.mm.1 |
TC0500002073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018830.mm.1 |
NM_013569.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018830.mm.1 |
NM_013569.2 |
Refseq |