Assay Details
| Gene Symbol: Mina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025910.3 | NP_080186.3 | 1 - 2 | 167 | 76 | |
| GenBank mRNA | AK007810.1 | - | 1 - 2 | 137 | 76 | |
| AK013451.1 | - | 1 - 2 | 127 | 76 | ||
| AK014426.1 | - | 1 - 2 | 121 | 76 | ||
| AK031671.1 | - | 1 - 2 | 101 | 76 | ||
| AK032595.1 | - | 1 - 2 | 138 | 76 | ||
| BC025109.1 | - | 1 - 2 | 89 | 4236994 | 76 | |
| BC058242.1 | - | 1 - 2 | 76 | 6308890 | 76 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK007810.1,AK013451.1,AK031671.1,AK032595.1,BC025109.1,BC058242.1,NM_025910.3 transcripts.
Set Membership: |
|

Mina
Genomic Map