Assay Details
Gene Symbol: Psmd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026000.2 | NP_080276.2 | 3 - 4 | 568 | 146 | |
GenBank mRNA | AK002280.1 | - | 3 - 4 | 492 | 146 | |
AK010276.1 | - | 3 - 4 | 456 | 146 | ||
AK011984.1 | - | 3 - 4 | 513 | 146 | ||
AK145487.1 | - | 3 - 4 | 513 | 146 | ||
AK168339.1 | - | 3 - 4 | 491 | 146 | ||
AK168863.1 | - | 3 - 4 | 510 | 146 | ||
BC051930.1 | - | 3 - 4 | 453 | 6403128 | 146 | |
BC060245.1 | - | 3 - 4 | 496 | 6819088 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
